missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pentyn-2-ol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0275214

 View more versions of this product

Catalog No. AAL0275214

Explore available promotions

Description

Description

4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

Solubility
Miscible with chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications

Specifications

4-Pentyn-2-ol
0.892
37°C (98°F)
1.438
25 g
1734245
Miscible with chloroform and ethyl acetate.
CC(CC#C)O
84.12
84.12
2117-11-5
126°C to 127°C
C5H8O
MFCD00004555
UN1987
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
JTHLRRZARWSHBE-UHFFFAOYNA-N
pent-4-yn-2-ol
92915
97%

Chemical Identifiers

2117-11-5
MFCD00004555
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 218-321-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only