missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pentyn-2-ol, 97%, Thermo Scientific Chemicals

$92.22 - $295.84

Chemical Identifiers

CAS 2117-11-5
MDL Number MFCD00004555
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0275206
View Documents
Thermo Scientific Chemicals
L0275206
5 g
Each for $92.22
 
AAL0275214
View Documents
Thermo Scientific Chemicals
L0275214
25 g
Each for $295.84
 
Description

Description

4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

Solubility
Miscible with chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications

Chemical Identifiers

2117-11-5
MFCD00004555

Specifications

2117-11-5
126°C to 127°C
C7H14OSi
MFCD00004555
UN1987
4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
VSTMVAFCUIGHQI-UHFFFAOYSA-N
4-trimethylsilylbut-3-yn-1-ol
2760827
97%
0.892
37°C (98°F)
1.438
5 g
1734245
Miscible with chloroform and ethyl acetate.
C[Si](C)(C)C#CCCO
142.27
84.12
4-Pentyn-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 218-321-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only