missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pentyn-2-ol, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 392670050

 View more versions of this product

Catalog No. AC392670050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Pentyn-2-ol
97.0
0.8960g/mL
37°C
97% min. (GC)
C5H8O
CHCCH2CH(OH)CH3
5g
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
CC(CC#C)O
84.12
84.12
2117-11-5
100.0
126°C to 127°C
Authentic
Glass bottle
1.4360 to 1.438
MFCD00004555
0.896
JTHLRRZARWSHBE-UHFFFAOYNA-N
pent-4-yn-2-ol
92915
97+%

Chemical Identifiers

2117-11-5
84.12
JTHLRRZARWSHBE-UHFFFAOYNA-N
92915
CC(CC#C)O
C5H8O
MFCD00004555
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
pent-4-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be con

GHS Signal Word: Warning

EINECSNumber : 218-321-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only