missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pentyn-2-ol, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 2117-11-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00004555
InChI Key JTHLRRZARWSHBE-UHFFFAOYNA-N
Synonym 4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
PubChem CID 92915
IUPAC Name pent-4-yn-2-ol
SMILES CC(CC#C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC392670010
SDS View Documents
Thermo Scientific Chemicals
392670010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC392670050
SDS View Documents
Thermo Scientific Chemicals
392670050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC392670250
SDS View Documents
Thermo Scientific Chemicals
392670250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2117-11-5
84.12
JTHLRRZARWSHBE-UHFFFAOYNA-N
92915
CC(CC#C)O
C5H8O
MFCD00004555
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
pent-4-yn-2-ol

Specifications

2117-11-5
100.0
126°C to 127°C
Authentic
Glass bottle
1.4360 to 1.438
MFCD00004555
0.896
JTHLRRZARWSHBE-UHFFFAOYNA-N
pent-4-yn-2-ol
92915
97+%
97.0
0.8960g/mL
37°C
97% min. (GC)
C5H8O
CHCCH2CH(OH)CH3
1g
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
CC(CC#C)O
84.12
84.12
4-Pentyn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-321-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only