Learn More
4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals
$194.94 - $194.94
Chemical Identifiers
CAS | 16520-62-0 |
---|---|
Molecular Formula | C10H10 |
Molecular Weight (g/mol) | 130.19 |
InChI Key | QDEOKXOYHYUKMS-UHFFFAOYSA-N |
Synonym | 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne |
PubChem CID | 123360 |
IUPAC Name | but-3-ynylbenzene |
SMILES | C#CCCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC449770050
|
Thermo Scientific Chemicals
449770050 |
5 g |
Each for $194.94
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16520-62-0 | |
130.19 | |
4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne | |
but-3-ynylbenzene |
C10H10 | |
QDEOKXOYHYUKMS-UHFFFAOYSA-N | |
123360 | |
C#CCCC1=CC=CC=C1 |
Specifications
16520-62-0 | |
190.0°C | |
96% min. (GC) | |
5 g | |
4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne | |
QDEOKXOYHYUKMS-UHFFFAOYSA-N | |
but-3-ynylbenzene | |
123360 | |
97% |
0.9260g/mL | |
66°C | |
C10H10 | |
0.926 | |
Solubility in water: insoluble | |
C#CCCC1=CC=CC=C1 | |
130.19 | |
130.19 | |
4-Phenyl-1-butyne |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only