missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylcyclohexanone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107911000

 View more versions of this product

Catalog No. AC107911000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-tert-Butylcyclohexanone
98.5
48.0°C to 51.0°C
113.0°C to 116.0°C (20.0 mmHg)
Authentic
Plastic bottle
(CH3)3CC6H9(=O)
100 g
4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
YKFKEYKJGVSEIX-UHFFFAOYSA-N
4-tert-butylcyclohexan-1-one
7392
99%
98-53-3
100.0
White
96°C
98.5% min. (GC)
C10H18O
MFCD00001642
07, I, 29
Solubility in water: insoluble
CC(C)(C)C1CCC(=O)CC1
154.25
154.25
Crystalline Powder

Chemical Identifiers

98-53-3
154.25
YKFKEYKJGVSEIX-UHFFFAOYSA-N
7392
CC(C)(C)C1CCC(=O)CC1
C10H18O
MFCD00001642
4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
4-tert-butylcyclohexan-1-one
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 202-678-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only