missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylcyclohexanone, 99%, Thermo Scientific Chemicals

$132.18 - $132.18

Chemical Identifiers

CAS 98-53-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001642
InChI Key YKFKEYKJGVSEIX-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
PubChem CID 7392
IUPAC Name 4-tert-butylcyclohexan-1-one
SMILES CC(C)(C)C1CCC(=O)CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC107911000
SDS View Documents
Thermo Scientific Chemicals
107911000
100 g Plastic bottle
Each for $132.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-53-3
154.25
YKFKEYKJGVSEIX-UHFFFAOYSA-N
7392
CC(C)(C)C1CCC(=O)CC1
C10H18O
MFCD00001642
4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
4-tert-butylcyclohexan-1-one

Specifications

98-53-3
100.0
White
96°C
98.5% min. (GC)
C10H18O
MFCD00001642
07, I, 29
Solubility in water: insoluble
CC(C)(C)C1CCC(=O)CC1
154.25
154.25
Crystalline Powder
98.5
48.0°C to 51.0°C
113.0°C to 116.0°C (20.0 mmHg)
Authentic
Plastic bottle
(CH3)3CC6H9(=O)
100 g
4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
YKFKEYKJGVSEIX-UHFFFAOYSA-N
4-tert-butylcyclohexan-1-one
7392
99%
4-tert-Butylcyclohexanone
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 202-678-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only