missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylphenylacetylene, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04392004

 View more versions of this product

Catalog No. AA4392004


Explore available promotions

Description

Description

4-tert-Butylphenylacetylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-tert-Butylphenylacetylene
0.878 g/mL
82°C (180°F)
C12H14
MFCD00190197
UN3082
Difficult to mix with water.
CC(C)(C)C1=CC=C(C=C1)C#C
158.244
158.24
Liquid
772-38-3
130°C (40 mmHg)
Characteristic
1.5297
2 g
1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
ZSYQVVKVKBVHIL-UHFFFAOYSA-N
1-tert-butyl-4-ethynylbenzene
2757302
≥90%

Chemical Identifiers

772-38-3
158.244
ZSYQVVKVKBVHIL-UHFFFAOYSA-N
2757302
CC(C)(C)C1=CC=C(C=C1)C#C
C12H14
MFCD00190197
1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
1-tert-butyl-4-ethynylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only