Learn More
4-tert-Butylphenylacetylene, 90+%, Thermo Scientific Chemicals
$63.29 - $219.26
Chemical Identifiers
CAS | 772-38-3 |
---|---|
Molecular Formula | C12H14 |
Molecular Weight (g/mol) | 158.244 |
MDL Number | MFCD00190197 |
InChI Key | ZSYQVVKVKBVHIL-UHFFFAOYSA-N |
Synonym | 1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene |
PubChem CID | 2757302 |
IUPAC Name | 1-tert-butyl-4-ethynylbenzene |
SMILES | CC(C)(C)C1=CC=C(C=C1)C#C |
Description
4-tert-Butylphenylacetylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
772-38-3 | |
158.244 | |
ZSYQVVKVKBVHIL-UHFFFAOYSA-N | |
2757302 | |
CC(C)(C)C1=CC=C(C=C1)C#C |
C12H14 | |
MFCD00190197 | |
1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene | |
1-tert-butyl-4-ethynylbenzene |
Specifications
772-38-3 | |
130°C (40 mmHg) | |
Characteristic | |
1.5297 | |
2 g | |
1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene | |
ZSYQVVKVKBVHIL-UHFFFAOYSA-N | |
1-tert-butyl-4-ethynylbenzene | |
2757302 | |
≥90% | |
4-tert-Butylphenylacetylene |
0.878 g/mL | |
82°C (180°F) | |
C12H14 | |
MFCD00190197 | |
UN3082 | |
Difficult to mix with water. | |
CC(C)(C)C1=CC=C(C=C1)C#C | |
158.244 | |
158.24 | |
Liquid |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only