missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 445570010

 View more versions of this product

Catalog No. AC445570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-(Tetrazol-5-yl)phenylboronic acid
95% min. (HPLC)
C7H7BN4O2
4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
B(C1=CC=C(C=C1)C2=NNN=N2)(O)O
189.97
189.97
179942-55-3
Glass Bottle
1g
DXUPJOQUAAVAGV-UHFFFAOYSA-N
[4-(2H-tetrazol-5-yl)phenyl]boronic acid
46737995
97%

Chemical Identifiers

179942-55-3
189.97
4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
[4-(2H-tetrazol-5-yl)phenyl]boronic acid
C7H7BN4O2
DXUPJOQUAAVAGV-UHFFFAOYSA-N
46737995
B(C1=CC=C(C=C1)C2=NNN=N2)(O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only