missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 179942-55-3
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
InChI Key DXUPJOQUAAVAGV-UHFFFAOYSA-N
Synonym 4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
PubChem CID 46737995
IUPAC Name [4-(2H-tetrazol-5-yl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C2=NNN=N2)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445570010
SDS View Documents
Thermo Scientific Chemicals
445570010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445570050
SDS View Documents
Thermo Scientific Chemicals
445570050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

179942-55-3
189.97
4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
[4-(2H-tetrazol-5-yl)phenyl]boronic acid
C7H7BN4O2
DXUPJOQUAAVAGV-UHFFFAOYSA-N
46737995
B(C1=CC=C(C=C1)C2=NNN=N2)(O)O

Specifications

179942-55-3
Glass Bottle
1g
DXUPJOQUAAVAGV-UHFFFAOYSA-N
[4-(2H-tetrazol-5-yl)phenyl]boronic acid
46737995
97%
95% min. (HPLC)
C7H7BN4O2
4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
B(C1=CC=C(C=C1)C2=NNN=N2)(O)O
189.97
189.97
4-(Tetrazol-5-yl)phenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only