missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2618006

 View more versions of this product

Catalog No. AAH2618006


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Trifluoromethoxy)phenylhydrazine hydrochloride
∼225°C (decomposition)
MFCD00053033
4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride
Cl.NNC1=CC=C(OC(F)(F)F)C=C1
228.60
228.6
133115-72-7
C7H8ClF3N2O
5 g
KQXZVSQCMVKMBK-UHFFFAOYSA-N
[4-(trifluoromethoxy)phenyl]hydrazine;hydrochloride
2777328
98%

Chemical Identifiers

133115-72-7
228.60
KQXZVSQCMVKMBK-UHFFFAOYSA-N
2777328
Cl.NNC1=CC=C(OC(F)(F)F)C=C1
C7H8ClF3N2O
MFCD00053033
4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride
[4-(trifluoromethoxy)phenyl]hydrazine;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only