Learn More
4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
$93.92 - $1,001.05
Chemical Identifiers
CAS | 133115-72-7 |
---|---|
Molecular Formula | C7H8ClF3N2O |
Molecular Weight (g/mol) | 228.60 |
MDL Number | MFCD00053033 |
InChI Key | KQXZVSQCMVKMBK-UHFFFAOYSA-N |
Synonym | 4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride |
PubChem CID | 2777328 |
IUPAC Name | [4-(trifluoromethoxy)phenyl]hydrazine;hydrochloride |
SMILES | Cl.NNC1=CC=C(OC(F)(F)F)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2618003
|
Thermo Scientific Chemicals
H2618003 |
1 g |
N/A
|
|
|||||
AAH2618006
|
Thermo Scientific Chemicals
H2618006 |
5 g |
N/A
|
|
|||||
AAH2618014
|
Thermo Scientific Chemicals
H2618014 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
133115-72-7 | |
228.60 | |
KQXZVSQCMVKMBK-UHFFFAOYSA-N | |
2777328 | |
Cl.NNC1=CC=C(OC(F)(F)F)C=C1 |
C7H8ClF3N2O | |
MFCD00053033 | |
4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride | |
[4-(trifluoromethoxy)phenyl]hydrazine;hydrochloride |
Specifications
133115-72-7 | |
C7H8ClF3N2O | |
1 g | |
KQXZVSQCMVKMBK-UHFFFAOYSA-N | |
[4-(trifluoromethoxy)phenyl]hydrazine;hydrochloride | |
2777328 | |
98% |
∼225°C (decomposition) | |
MFCD00053033 | |
4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride | |
Cl.NNC1=CC=C(OC(F)(F)F)C=C1 | |
228.60 | |
228.6 | |
4-(Trifluoromethoxy)phenylhydrazine hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only