Learn More
4-(Trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2237403
Description
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.
Solubility
Soluble in DMSO, not in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
4-(Trifluoromethyl)benzeneboronic acid | |
238°C to 242°C | |
MFCD00151855 | |
3544189 | |
Soluble in DMSO,not in water | |
OB(O)C1=CC=C(C=C1)C(F)(F)F | |
189.93 | |
189.93 |
128796-39-4 | |
C7H6BF3O2 | |
1 g | |
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid | |
ALMFIOZYDASRRC-UHFFFAOYSA-N | |
[4-(trifluoromethyl)phenyl]boronic acid | |
2734389 | |
98% |
Chemical Identifiers
128796-39-4 | |
189.93 | |
ALMFIOZYDASRRC-UHFFFAOYSA-N | |
2734389 | |
OB(O)C1=CC=C(C=C1)C(F)(F)F |
C7H6BF3O2 | |
MFCD00151855 | |
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid | |
[4-(trifluoromethyl)phenyl]boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only