missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

$140.52 - $1727.75

Chemical Identifiers

CAS 128796-39-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151855
InChI Key ALMFIOZYDASRRC-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
PubChem CID 2734389
IUPAC Name [4-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2237403
View Documents
Thermo Scientific Chemicals
B2237403
1 g
Each for $140.52
 
AAB2237406
View Documents
Thermo Scientific Chemicals
B2237406
5 g
Each for $416.03
 
AAB2237414
View Documents
Thermo Scientific Chemicals
B2237414
25 g
Each for $1,727.75
 
Description

Description

4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

Solubility
Soluble in DMSO, not in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

128796-39-4
189.93
ALMFIOZYDASRRC-UHFFFAOYSA-N
2734389
OB(O)C1=CC=C(C=C1)C(F)(F)F
C7H6BF3O2
MFCD00151855
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
[4-(trifluoromethyl)phenyl]boronic acid

Specifications

128796-39-4
C7H6BF3O2
1 g
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
ALMFIOZYDASRRC-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]boronic acid
2734389
98%
238°C to 242°C
MFCD00151855
3544189
Soluble in DMSO,not in water
OB(O)C1=CC=C(C=C1)C(F)(F)F
189.93
189.93
4-(Trifluoromethyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only