missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1110714

 View more versions of this product

Catalog No. AAA1110714


Explore available promotions

Description

Description

It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in dichloromethane and methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

4-Vinylbenzoic acid
∼142°C
C9H8O2
25 g
Light sensitive
Soluble in dichloromethane and methanol
C=CC1=CC=C(C=C1)C(=O)O
148.161
148.16
1075-49-6
1.3
MFCD00002569
2041130
4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
IRQWEODKXLDORP-UHFFFAOYSA-N
4-ethenylbenzoic acid
14098
98%

Chemical Identifiers

1075-49-6
148.161
IRQWEODKXLDORP-UHFFFAOYSA-N
14098
C=CC1=CC=C(C=C1)C(=O)O
C9H8O2
MFCD00002569
4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
4-ethenylbenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-053-4

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only