Learn More
4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals
$206.10 - $2282.38
Chemical Identifiers
CAS | 1075-49-6 |
---|---|
Molecular Formula | C9H8O2 |
Molecular Weight (g/mol) | 148.161 |
MDL Number | MFCD00002569 |
InChI Key | IRQWEODKXLDORP-UHFFFAOYSA-N |
Synonym | 4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624 |
PubChem CID | 14098 |
IUPAC Name | 4-ethenylbenzoic acid |
SMILES | C=CC1=CC=C(C=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1110703
|
Thermo Scientific Chemicals
A1110703 |
1 g |
Each for $206.10
|
|
|||||
AAA1110706
|
Thermo Scientific Chemicals
A1110706 |
5 g |
Each for $682.64
|
|
|||||
AAA1110714
|
Thermo Scientific Chemicals
A1110714 |
25 g |
Each for $2,282.38
|
|
|||||
Description
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in dichloromethane and methanol
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
1075-49-6 | |
148.161 | |
IRQWEODKXLDORP-UHFFFAOYSA-N | |
14098 | |
C=CC1=CC=C(C=C1)C(=O)O |
C9H8O2 | |
MFCD00002569 | |
4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624 | |
4-ethenylbenzoic acid |
Specifications
1075-49-6 | |
1.3 | |
MFCD00002569 | |
2041130 | |
4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624 | |
IRQWEODKXLDORP-UHFFFAOYSA-N | |
4-ethenylbenzoic acid | |
14098 | |
98% |
∼142°C | |
C9H8O2 | |
1 g | |
Light sensitive | |
Soluble in dichloromethane and methanol | |
C=CC1=CC=C(C=C1)C(=O)O | |
148.161 | |
148.16 | |
4-Vinylbenzoic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-053-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only