National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Arc Flash Coat
National Safety Apparel™ arc flash coats come with hook and loop front closure with drawstring waist
Supplier:  National Safety Apparel™ C04NPQHLTD3X32
 View more versions of this product
Catalog No. 17001218
Explore available promotions
Description
Description
  • 50% lighter tha standard 40 cal/cm2 arc flash kits
  • Stand up collar for extra coverage, can be turned down
  • Nomex knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • FR fabric will not melt, drip, or ignite
  • Available in a convenient kit
Specifications
Specifications
3X-Large
Olive Green
FR Hook and Loop with Buckle
40 cal/cm²
RevoLite™
Open
Stitched
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269