Learn More
Agilent BioTek 405 TS Touch Microplate Washer
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications.
Supplier: Agilent BioTek 405TSUVSQSN
Description
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.
The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Specifications
ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes | |
USB (2) | |
<3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) | |
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates | |
96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s | |
Microplate Washer | |
1 to 250 | |
LED | |
Internal positive displacement pump | |
50/60 Hz | |
Automated Internal 4-Buffer Switching; Ultrasonic Advantage; Optimized Cell Washing; 96-/384-well microplate washing; Verify technology | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
With internal buffer switching - 36 lb.; Without - 30 lb. |
CE and TUV marked. RoHS compliant. | |
Optional (4 or 10 L) | |
25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) | |
≤2 μL/well | |
Programmable in minutes and seconds, up to 60 min. | |
With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
25 to 3000 μL/well | |
High to Low Flow; Optimized rates for cell assays | |
ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays | |
405TSUVSQ | |
14 in. | |
10 in. | |
17 in. | |
High Resolution LED Backlit Touch Screen | |
100/240 V |