Learn More
Agilent BioTek 405 TS Touch Microplate Washer
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications.
$19387.75 - $43933.18
Specifications
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
---|---|
Connections | USB (2) |
Bottles | Optional (4 or 10 L) |
Dispense Volume | 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) |
Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates |
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
BT405TSR
|
Agilent BioTek
405TSRSN |
405TSR | 96-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSRS
|
Agilent BioTek
405TSRSSN |
405TSRS | 96-well microplate washing, Ultrasonic Advantage |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSRV
|
Agilent BioTek
405TSRVSN |
405TSRV | Automated Internal 4-Buffer Switching; 96-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSRVS
|
Agilent BioTek
405TSRVSSN |
405TSRVS | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSHT
|
Agilent BioTek
405TSHTSN |
405TSHT | Ultrasonic Advantage; Faster 384-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSU
|
Agilent BioTek
405TSUSN |
405TSU | Optimized Cell Washing; 96-/384-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSUS
|
Agilent BioTek
405TSUSSN |
405TSUS | Optimized Cell Washing; Ultrasonic Advantage; 96-/384-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSUV
|
Agilent BioTek
405TSUVSN |
405TSUV | Automated Internal 4-Buffer Switching; 96-/384-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSUVS
|
Agilent BioTek
405TSUVSSN |
405TSUVS | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; Optimized Cell Washing; 96-/384-well microplate washing |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSRSQ
|
Agilent BioTek
405TSRSQSN |
405TSRSQ | Ultrasonic Advantage; 96-well microplate washing; Verify Technology |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSRVSQ
|
Agilent BioTek
405TSRVSQSN |
405TSRVSQ | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-well microplate washing; Verify Technology |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSUSQ
|
Agilent BioTek
405TSUSQSN |
405TSUSQ | Optimized Cell Washing; Ultrasonic Advantage; 96-/384-well microplate washing; Verify technology |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405TSUVSQ
|
Agilent BioTek
405TSUVSQSN |
405TSUVSQ | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; Optimized Cell Washing; 96-/384-well microplate washing; Verify technology |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.
The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Specifications
CE and TUV marked. RoHS compliant. | |
Optional (4 or 10 L) | |
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates | |
Programmable in minutes and seconds, up to 60 min. | |
With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
25 to 3000 μL/well | |
Internal positive displacement pump | |
14 in. | |
10 in. | |
17 in. | |
High Resolution LED Backlit Touch Screen | |
100/240 V |
USB (2) | |
25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) | |
≤2 μL/well | |
Microplate Washer | |
1 to 250 | |
LED | |
50/60 Hz | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
With internal buffer switching - 36 lb.; Without - 30 lb. |