missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Non-Combustible Outdoor Safety Lockers

Designed for storage of flammable and combustible liquids contained in chemical drums varying in capacities

Supplier:  Justrite™ 911090

 View more versions of this product

Catalog No. 19-414-011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Made of heavy gauge steel (backed by 15 year structural warranty)
  • Leakproof all welded sump secondary containment
  • Removable, galvanized steel grating
  • Three-point keyed locking hardware
  • Natural draft ventilation 
  • Multiple static grounding connections 
  • Anchoring plates for maximum hold down protection 
  • Chemical and UV resistant coating, inside and out 
  • Forklift channels for easy relocation
  • Available with and without Explosion Relief Panels
  • NFPA 704 diamond decal provides "Right to Know" information
  • FM-approved - including sump capacity and wind/snow load requirements
Model Specific Features:
  • With Explosion relief panels designed to release at 20 psf (911021, 911041, 911061, 911091, 911121, and 911161)
  • Without Explosion relief panels (911020, 911040, 911060, 911090, 911120 and 911160)

Certifications

FM approved

Compliance

Meets EPA, NFPA 30 and NFPA 1 requirements

Specifications

Specifications

84 in.
Heavy gauge steel
Hazardous/chemical storage
198.1 cm
91 in.
No
86 in.
198.1 x 198.1 x 213.4 cm (78 x 78 x 84 in.)
218.4 cm
9 x 55 gal. drums
1
78 in.
213.4 cm
Non-Combustible Outdoor Safety Locker
198.1 cm
97 in.
Steel
231.1 cm
78 in.
Bone
246.4 cm
Three-point lock
FM approved, EPA, NFPA 30, NFPA 1 compliant
Videos
SDS
Documents

Documents

Promotions

Promotions