missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Non-Combustible Outdoor Safety Lockers
Designed for storage of flammable and combustible liquids contained in chemical drums varying in capacities
$25416.42 - $73476.59
Specifications
Certifications/Compliance | FM approved, EPA, NFPA 30, NFPA 1 compliant |
---|---|
Description | Heavy gauge steel |
Color | Bone |
Material | Steel |
Product Type | Non-Combustible Outdoor Safety Locker |
Catalog Number | Mfr. No. | Load Bearing Capacity | Explosion Relief Panel | Dimensions (D x W x H) Interior | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Load Bearing Capacity | Explosion Relief Panel | Dimensions (D x W x H) Interior | Price | Quantity | |||
19-414-005
|
Justrite™
911020 |
2 x 55 gal. drums | No | 76.2 x 137.2 x 141 cm (30 x 54 x 55.5 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-006
|
Justrite™
911021 |
2 x 55 gal. drums | Yes | 76.2 x 137.2 x 141 cm (30 x 54 x 55.5 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-007
|
Justrite™
911040 |
4 x 55 gal. drums | No | 137.2 x 137.2 x 157.5 cm (54 x 54 x 62 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-008
|
Justrite™
911041 |
4 x 55 gal. drums | Yes | 137.2 x 137.2 x 157.5 cm (54 x 54 x 62 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-009
|
Justrite™
911060 |
6 x 55 gal. drums | No | 137.2 x 198.1 x 213.4 cm (54 x 78 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-010
|
Justrite™
911061 |
6 x 55 gal. drums | Yes | 137.2 x 198.1 x 213.4 cm (54 x 78 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-011
|
Justrite™
911090 |
9 x 55 gal. drums | No | 198.1 x 198.1 x 213.4 cm (78 x 78 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-012
|
Justrite™
911091 |
9 x 55 gal. drums | Yes | 198.1 x 198.1 x 213.4 cm (78 x 78 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-013
|
Justrite™
911120 |
12 x 55 gal. drums | No | 259.1 x 198.1 x 234.1 cm (102 x 78 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-014
|
Justrite™
911121 |
12 x 55 gal. drums | Yes | 259.1 x 198.1 x 213.4 cm (102 x 78 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-015
|
Justrite™
911160 |
16 x 55 gal. drums | No | 259.1 x 259.1 x 213.4 cm (102 x 102 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19-414-016
|
Justrite™
911161 |
16 x 55 gal. drums | Yes | 259.1 x 259.1 x 213.4 cm (102 x 102 x 84 in.) |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Made of heavy gauge steel (backed by 15 year structural warranty)
- Leakproof all welded sump secondary containment
- Removable, galvanized steel grating
- Three-point keyed locking hardware
- Natural draft ventilation
- Multiple static grounding connections
- Anchoring plates for maximum hold down protection
- Chemical and UV resistant coating, inside and out
- Forklift channels for easy relocation
- Available with and without Explosion Relief Panels
- NFPA 704 diamond decal provides“Right to Know”information
- FM-approved - including sump capacity and wind/snow load requirements
- With Explosion relief panels designed to release at 20 psf (911021, 911041, 911061, 911091, 911121, and 911161)
- Without Explosion relief panels (911020, 911040, 911060, 911090, 911120 and 911160)
Certifications
FM approved
Compliance
Meets EPA, NFPA 30 and NFPA 1 requirements
Specifications
FM approved, EPA, NFPA 30, NFPA 1 compliant | |
Bone | |
Non-Combustible Outdoor Safety Locker | |
Hazardous/chemical storage |
Heavy gauge steel | |
Steel | |
Three-point lock | |
1 |