missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Non-Combustible Outdoor Safety Lockers
Designed for storage of flammable and combustible liquids contained in chemical drums varying in capacities
Supplier: Justrite™ 911091
Description
- Made of heavy gauge steel (backed by 15 year structural warranty)
- Leakproof all welded sump secondary containment
- Removable, galvanized steel grating
- Three-point keyed locking hardware
- Natural draft ventilation
- Multiple static grounding connections
- Anchoring plates for maximum hold down protection
- Chemical and UV resistant coating, inside and out
- Forklift channels for easy relocation
- Available with and without Explosion Relief Panels
- NFPA 704 diamond decal provides“Right to Know”information
- FM-approved - including sump capacity and wind/snow load requirements
- With Explosion relief panels designed to release at 20 psf (911021, 911041, 911061, 911091, 911121, and 911161)
- Without Explosion relief panels (911020, 911040, 911060, 911090, 911120 and 911160)
Certifications
FM approved
Compliance
Meets EPA, NFPA 30 and NFPA 1 requirements
Specifications
84 in. | |
Heavy gauge steel | |
Hazardous/chemical storage | |
198.1 cm | |
91 in. | |
Yes | |
86 in. | |
198.1 x 198.1 x 213.4 cm (78 x 78 x 84 in.) | |
218.4 cm | |
9 x 55 gal. drums | |
1 | |
78 in. |
213.4 cm | |
Non-Combustible Outdoor Safety Locker | |
198.1 cm | |
97 in. | |
Steel | |
231.1 cm | |
78 in. | |
Bone | |
246.4 cm | |
Three-point lock | |
FM approved, EPA, NFPA 30, NFPA 1 compliant |