Brady™ BASIC Oil Only Absorbent Pads
For cleaning up petroleum based spills and leaks
Supplier:  Brady™ BPO100
 View more versions of this product
Catalog No. 17-111-120
Explore available promotions
Description
Description
  • Absorbs oil and repels liquid
  • Single-ply Oil Only meltblown polypropylene made
  • Applied during emergency spills for fast and easy clean-up
  • Available in various sizes
Specifications
Specifications
White
Absorbent Pad, Oil-only
100/Cs.
38.1 x 43.1 cm (15 x 17 in.)
20.5 gal./Bale
BASIC Series
Heavyweight