missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ BASIC Oil Only Absorbent Pads
For cleaning up petroleum based spills and leaks
Supplier: Brady™ BPO500
Description
Specifications
White | |
Polypropylene | |
BASIC Series | |
Lightweight |
17 gal./Bale | |
Absorbent Pad, Oil-only | |
100/Cs. | |
38.1 x 43.1 cm (15 x 17 in.) |