DuPont™ Tychem™ 6000 Taped Seam Coveralls With Attached Socks (TAA Compliant)
Widely used by military personnel and first responders for chemical warfare situations
Supplier: DuPont™ TF169TGYMD0006TV
Description
- Strong and durable coverall with broad chemical barrier
- Tychem™ 6000 coverall is made of barrier film laminated to Tyvek™ substrate
- For use when potential exposure to industrial chemicals and chemical warfare agents exists
- Provides protection against over 180 chemical challenges for at least 30 minutes
- Available in low-visibility gray fabric for discretionary purposes
- With taped seams and attached hood (respirator fit)
- Front zipper closure and storm flap with tape closure
- Elastic wrists and elastic ankles
Compliance
TAA Compliant
Specifications
Medium | |
Tychem | |
Gray | |
Taped |