DuPont™ Tychem™ 6000 Taped Seam Coveralls With Attached Socks (TAA Compliant)
Widely used by military personnel and first responders for chemical warfare situations
$557.42 - $730.58
Specifications
Material Tychem
Color Gray
Seam Type Taped
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-162-817
View Documents
DuPont™
TF169TGY4X0006TV
4X-Large
Case of 6 Each for $676.40
19-162-820
View Documents
DuPont™
TF169TGYLG0006TV
Large
Case of 6 Each for $557.42
19-162-816
View Documents
DuPont™
TF169TGY3X0006TV
3X-Large
Case of 6 Each for $626.33
19-162-821
View Documents
DuPont™
TF169TGYMD0006TV
Medium
Case of 6 Each for $557.42
19-162-819
View Documents
DuPont™
TF169TGY5X0006TV
5X-Large
Case of 6 Each for $730.58
19-162-822
View Documents
DuPont™
TF169TGYSM0006TV
Small
Case of 6 Each for $557.42
Description
Description
  • Strong and durable coverall with broad chemical barrier
  • Tychem™ 6000 coverall is made of barrier film laminated to Tyvek™ substrate
  • For use when potential exposure to industrial chemicals and chemical warfare agents exists
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Available in low-visibility gray fabric for discretionary purposes
  • With taped seams and attached hood (respirator fit)
  • Front zipper closure and storm flap with tape closure
  • Elastic wrists and elastic ankles
Compliance
TAA Compliant
Specifications
Specifications
Tychem
Gray
Taped
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'