missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tychem™ CPF 3 Aprons

For rigorous activities and rugged situations involving liquid splash

Supplier:  DuPont™ C3275TTNMD000600

 View more versions of this product

Catalog No. 19-162-803


Explore available promotions

Description

Description

  • Tychem™ CPF 3 is multi-layer barrier film laminated to durable 2.0 oz./yd2 polypropylene fabric
  • Strong and durable 44 in. long apron with broad chemical barrier
  • Tychem CPF 3 provides at least 30 minutes of protection against 149 chemical challenges
  • Taped seams and bound snap neck
  • Apron with attached long sleeves, waist ties and elastic wrists
  • Tan color for discretionary purposes
  • Tychem CPF 3 198 T/199 T are certified to NFPA 1992 (2005 Edition)

Specifications

Specifications

Medium
Tan
111.7 cm
Tychem
44 in.
Taped
Videos
SDS
Documents

Documents

Promotions

Promotions