missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(4-Nitrophenyl)-2-furaldehyde 98.0+%, TCI America™
Supplier: TCI America N082025G
Specifications
5-(4-Nitrophenyl)-2-furaldehyde | |
207°C | |
MFCD00124191 | |
5-4-nitrophenyl-2-furaldehyde, 5-4-nitrophenyl furan-2-carbaldehyde, 5-4-nitrophenyl-2-furancarboxaldehyde, 5-4-nitrophenyl furfural, 5-4-nitro-phenyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 5-4-nitrophenyl, 5-p-nitrophenyl furfural, 5-4-nitrophenyl 2-furaldehyde, cambridge id 5143178, acmc-1bl92 | |
C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-] | |
217.18 | |
217.18 | |
Crystalline Powder |
7147-77-5 | |
C11H7NO4 | |
25 g | |
RTSOJVJDKNKNFU-UHFFFAOYSA-N | |
5-(4-nitrophenyl)furan-2-carbaldehyde | |
81567 | |
≥98.0% (GC) |
Chemical Identifiers
7147-77-5 | |
217.18 | |
RTSOJVJDKNKNFU-UHFFFAOYSA-N | |
81567 | |
C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-] |
C11H7NO4 | |
MFCD00124191 | |
5-4-nitrophenyl-2-furaldehyde, 5-4-nitrophenyl furan-2-carbaldehyde, 5-4-nitrophenyl-2-furancarboxaldehyde, 5-4-nitrophenyl furfural, 5-4-nitro-phenyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 5-4-nitrophenyl, 5-p-nitrophenyl furfural, 5-4-nitrophenyl 2-furaldehyde, cambridge id 5143178, acmc-1bl92 | |
5-(4-nitrophenyl)furan-2-carbaldehyde |
Safety and Handling
TSCA : No