missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(4-Nitrophenyl)-2-furaldehyde 98.0+%, TCI America™
$106.17 - $287.00
Chemical Identifiers
CAS | 7147-77-5 |
---|---|
Molecular Formula | C11H7NO4 |
Molecular Weight (g/mol) | 217.18 |
MDL Number | MFCD00124191 |
InChI Key | RTSOJVJDKNKNFU-UHFFFAOYSA-N |
Synonym | 5-4-nitrophenyl-2-furaldehyde, 5-4-nitrophenyl furan-2-carbaldehyde, 5-4-nitrophenyl-2-furancarboxaldehyde, 5-4-nitrophenyl furfural, 5-4-nitro-phenyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 5-4-nitrophenyl, 5-p-nitrophenyl furfural, 5-4-nitrophenyl 2-furaldehyde, cambridge id 5143178, acmc-1bl92 |
PubChem CID | 81567 |
IUPAC Name | 5-(4-nitrophenyl)furan-2-carbaldehyde |
SMILES | C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-] |
Chemical Identifiers
7147-77-5 | |
217.18 | |
RTSOJVJDKNKNFU-UHFFFAOYSA-N | |
81567 | |
C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-] |
C11H7NO4 | |
MFCD00124191 | |
5-4-nitrophenyl-2-furaldehyde, 5-4-nitrophenyl furan-2-carbaldehyde, 5-4-nitrophenyl-2-furancarboxaldehyde, 5-4-nitrophenyl furfural, 5-4-nitro-phenyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 5-4-nitrophenyl, 5-p-nitrophenyl furfural, 5-4-nitrophenyl 2-furaldehyde, cambridge id 5143178, acmc-1bl92 | |
5-(4-nitrophenyl)furan-2-carbaldehyde |
Specifications
7147-77-5 | |
C11H7NO4 | |
5 g | |
RTSOJVJDKNKNFU-UHFFFAOYSA-N | |
5-(4-nitrophenyl)furan-2-carbaldehyde | |
81567 | |
≥98.0% (GC) | |
5-(4-Nitrophenyl)-2-furaldehyde |
207°C | |
MFCD00124191 | |
5-4-nitrophenyl-2-furaldehyde, 5-4-nitrophenyl furan-2-carbaldehyde, 5-4-nitrophenyl-2-furancarboxaldehyde, 5-4-nitrophenyl furfural, 5-4-nitro-phenyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 5-4-nitrophenyl, 5-p-nitrophenyl furfural, 5-4-nitrophenyl 2-furaldehyde, cambridge id 5143178, acmc-1bl92 | |
C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-] | |
217.18 | |
217.18 | |
Crystalline Powder |
Safety and Handling
TSCA : No