Learn More
5,6,7,8-Tetrahydroquinoxaline, 99%, Thermo Scientific Chemicals
$49.16 - $101.16
Chemical Identifiers
CAS | 34413-35-9 |
---|---|
Molecular Formula | C8H10N2 |
Molecular Weight (g/mol) | 134.182 |
MDL Number | MFCD00038078 |
InChI Key | XCZPDOCRSYZOBI-UHFFFAOYSA-N |
Synonym | cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495 |
PubChem CID | 36822 |
IUPAC Name | 5,6,7,8-tetrahydroquinoxaline |
SMILES | C1CCC2=NC=CN=C2C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1120403
|
Thermo Scientific Chemicals
L1120403 |
1 g |
Each for $49.16
|
|
|||||
AAL1120406
|
Thermo Scientific Chemicals
L1120406 |
5 g |
Each for $101.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34413-35-9 | |
134.182 | |
XCZPDOCRSYZOBI-UHFFFAOYSA-N | |
36822 | |
C1CCC2=NC=CN=C2C1 |
C8H10N2 | |
MFCD00038078 | |
cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495 | |
5,6,7,8-tetrahydroquinoxaline |
Specifications
34413-35-9 | |
72°C to 74°C (4 mmHg) | |
C8H10N2 | |
MFCD00038078 | |
957053 | |
XCZPDOCRSYZOBI-UHFFFAOYSA-N | |
5,6,7,8-tetrahydroquinoxaline | |
36822 | |
99% |
1.061 | |
92°C (197°F) | |
1.542 | |
1 g | |
cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495 | |
C1CCC2=NC=CN=C2C1 | |
134.182 | |
134.18 | |
5,6,7,8-Tetrahydroquinoxaline |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 252-002-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only