Learn More
5-Acetoxymethyl-2-furaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 163400100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Acetoxymethyl-2-furaldehyde | |
10551-58-3 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00003233 | |
18, 15 | |
QAVITTVTXPZTSE-UHFFFAOYSA-N | |
(5-formylfuran-2-yl)methyl acetate | |
66349 | |
97% |
97% | |
96.0 | |
107°C | |
96% min. (GC) | |
C8H8O4 | |
10g | |
5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl | |
CC(=O)OCC1=CC=C(O1)C=O | |
168.15 | |
168.15 |
Chemical Identifiers
10551-58-3 | |
168.15 | |
QAVITTVTXPZTSE-UHFFFAOYSA-N | |
66349 | |
CC(=O)OCC1=CC=C(O1)C=O |
C8H8O4 | |
MFCD00003233 | |
5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl | |
(5-formylfuran-2-yl)methyl acetate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
EINECSNumber : 234-137-4
RUO â Research Use Only