missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Acetoxymethyl-2-furaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 10551-58-3
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00003233
InChI Key QAVITTVTXPZTSE-UHFFFAOYSA-N
Synonym 5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
PubChem CID 66349
IUPAC Name (5-formylfuran-2-yl)methyl acetate
SMILES CC(=O)OCC1=CC=C(O1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC163400020
SDS View Documents
Thermo Scientific Chemicals
163400020
2g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163400100
SDS View Documents Promotion Details
Thermo Scientific Chemicals
163400100
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

10551-58-3
168.15
QAVITTVTXPZTSE-UHFFFAOYSA-N
66349
CC(=O)OCC1=CC=C(O1)C=O
C8H8O4
MFCD00003233
5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
(5-formylfuran-2-yl)methyl acetate

Specifications

10551-58-3
100.0
Authentic
Glass bottle
MFCD00003233
18, 15
QAVITTVTXPZTSE-UHFFFAOYSA-N
(5-formylfuran-2-yl)methyl acetate
66349
97%
96.0
107°C
96% min. (GC)
C8H8O4
2g
5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
CC(=O)OCC1=CC=C(O1)C=O
168.15
168.15
5-Acetoxymethyl-2-furaldehyde, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

EINECSNumber : 234-137-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only