missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Acetylthiophene-2-boronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1522114

 View more versions of this product

Catalog No. AAL1522114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Acetylthiophene-2-boronic acid
142°C to 148°C
MFCD01075681
8681302
DCNMATSPQKWETQ-UHFFFAOYSA-N
(5-acetylthiophen-2-yl)boronic acid
4143377
98%
206551-43-1
C6H7BO3S
25 g
5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
B(C1=CC=C(S1)C(=O)C)(O)O
169.989
170

Chemical Identifiers

206551-43-1
169.989
DCNMATSPQKWETQ-UHFFFAOYSA-N
4143377
B(C1=CC=C(S1)C(=O)C)(O)O
C6H7BO3S
MFCD01075681
5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
(5-acetylthiophen-2-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only