Learn More
5-Acetylthiophene-2-boronic acid, 98%, Thermo Scientific Chemicals
$48.00 - $440.41
Chemical Identifiers
CAS | 206551-43-1 |
---|---|
Molecular Formula | C6H7BO3S |
Molecular Weight (g/mol) | 169.989 |
MDL Number | MFCD01075681 |
InChI Key | DCNMATSPQKWETQ-UHFFFAOYSA-N |
Synonym | 5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one |
PubChem CID | 4143377 |
IUPAC Name | (5-acetylthiophen-2-yl)boronic acid |
SMILES | B(C1=CC=C(S1)C(=O)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1522103
|
Thermo Scientific Chemicals
L1522103 |
1 g |
N/A
|
|
|||||
AAL1522106
|
Thermo Scientific Chemicals
L1522106 |
5 g |
N/A
|
|
|||||
AAL1522114
|
Thermo Scientific Chemicals
L1522114 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
206551-43-1 | |
169.989 | |
DCNMATSPQKWETQ-UHFFFAOYSA-N | |
4143377 | |
B(C1=CC=C(S1)C(=O)C)(O)O |
C6H7BO3S | |
MFCD01075681 | |
5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one | |
(5-acetylthiophen-2-yl)boronic acid |
Specifications
206551-43-1 | |
C6H7BO3S | |
1 g | |
5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one | |
B(C1=CC=C(S1)C(=O)C)(O)O | |
169.989 | |
170 | |
5-Acetylthiophene-2-boronic acid |
142°C to 148°C | |
MFCD01075681 | |
8681302 | |
DCNMATSPQKWETQ-UHFFFAOYSA-N | |
(5-acetylthiophen-2-yl)boronic acid | |
4143377 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only