missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aminoisophthalic Acid Hydrate 98.0+%, TCI America™

Supplier:  TCI America A129025G

 View more versions of this product

Catalog No. A129025G


Explore available promotions

Description

Description

Specifications

Specifications

5-Aminoisophthalic Acid Hydrate
White-Yellow
MFCD00002522
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
181.147
181.15
Crystalline Powder
99-31-0
C8H7NO4
25 g
KBZFDRWPMZESDI-UHFFFAOYSA-N
5-aminobenzene-1,3-dicarboxylic acid
66833
≥98.0% (T)

Chemical Identifiers

99-31-0
181.147
KBZFDRWPMZESDI-UHFFFAOYSA-N
66833
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
C8H7NO4
MFCD00002522
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
5-aminobenzene-1,3-dicarboxylic acid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions