missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aminoisophthalic Acid Hydrate 98.0+%, TCI America™

$61.22 - $456.22

Chemical Identifiers

CAS 99-31-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00002522
InChI Key KBZFDRWPMZESDI-UHFFFAOYSA-N
Synonym 5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
PubChem CID 66833
IUPAC Name 5-aminobenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)N)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A129025G
View Documents
TCI America
A129025G
25 g
Each for $61.22
 
A1290100G
View Documents
TCI America
A1290100G
100 g
Each for $159.97
 
A1290500G
View Documents
TCI America
A1290500G
500 g
Each for $456.22
 
Description

Description

Specifications

Chemical Identifiers

99-31-0
181.147
KBZFDRWPMZESDI-UHFFFAOYSA-N
66833
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
C8H7NO4
MFCD00002522
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
5-aminobenzene-1,3-dicarboxylic acid

Specifications

99-31-0
C8H7NO4
25 g
KBZFDRWPMZESDI-UHFFFAOYSA-N
5-aminobenzene-1,3-dicarboxylic acid
66833
≥98.0% (T)
5-Aminoisophthalic Acid Hydrate
White-Yellow
MFCD00002522
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
181.147
181.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'