Learn More
5-Bromo-1-pentene, 96%, Thermo Scientific Chemicals
$118.33 - $394.45
Chemical Identifiers
CAS | 1119-51-3 |
---|---|
Molecular Formula | C5H9Br |
Molecular Weight (g/mol) | 149.031 |
MDL Number | MFCD0000-364 |
InChI Key | LPNANKDXVBMDKE-UHFFFAOYSA-N |
Synonym | 5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide |
PubChem CID | 70704 |
IUPAC Name | 5-bromopent-1-ene |
SMILES | C=CCCCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1121906
|
Thermo Scientific Chemicals
A1121906 |
5 g |
Each for $118.33
|
|
||||||
AAA1121914
|
Thermo Scientific Chemicals
A1121914 |
25 g |
Each for $394.45
|
|
||||||
Description
5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.
Solubility
Immiscible with water.
Notes
Keep away from sources of ignition.
Chemical Identifiers
1119-51-3 | |
149.031 | |
LPNANKDXVBMDKE-UHFFFAOYSA-N | |
70704 | |
C=CCCCBr |
C5H9Br | |
MFCD0000-364 | |
5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide | |
5-bromopent-1-ene |
Specifications
1119-51-3 | |
126°C to 127°C | |
C5H9Br | |
Br(CH2)3CH=CH2 | |
5 g | |
506077 | |
Immiscible with water. | |
C=CCCCBr | |
149.031 | |
149.03 | |
5-Bromo-1-pentene |
1.259 | |
30°C (86°F) | |
1.4639 | |
MFCD0000-364 | |
UN1993 | |
5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide | |
LPNANKDXVBMDKE-UHFFFAOYSA-N | |
5-bromopent-1-ene | |
70704 | |
96% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 214-281-4
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only