5'-Bromo-2'-hydroxypropiophenone 98.0+%, TCI America™
Supplier:  TCI America B47691G
 View more versions of this product
Catalog No. B47691G
Explore available promotions
Description
Description
Specifications
Specifications
5′-Bromo-2′-hydroxypropiophenone
77°C
C9H9BrO2
1 g
CCC(=O)C1=CC(Br)=CC=C1O
229.07
229.07
Crystalline Powder
17764-93-1
156°C
MFCD00218577
NDXLYRKTHTZXRC-UHFFFAOYSA-N
1-(5-bromo-2-hydroxyphenyl)propan-1-one
736409
≥98.0% (GC,T)
Chemical Identifiers
17764-93-1
229.07
NDXLYRKTHTZXRC-UHFFFAOYSA-N
1-(5-bromo-2-hydroxyphenyl)propan-1-one
C9H9BrO2
MFCD00218577
736409
CCC(=O)C1=CC(Br)=CC=C1O
Videos
Safety and Handling
Safety and Handling
TSCA : No