missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5'-Bromo-2'-hydroxypropiophenone 98.0+%, TCI America™
$41.52 - $122.84
Chemical Identifiers
CAS | 17764-93-1 |
---|---|
Molecular Formula | C9H9BrO2 |
Molecular Weight (g/mol) | 229.07 |
MDL Number | MFCD00218577 |
InChI Key | NDXLYRKTHTZXRC-UHFFFAOYSA-N |
PubChem CID | 736409 |
IUPAC Name | 1-(5-bromo-2-hydroxyphenyl)propan-1-one |
SMILES | CCC(=O)C1=CC(Br)=CC=C1O |
Chemical Identifiers
17764-93-1 | |
229.07 | |
NDXLYRKTHTZXRC-UHFFFAOYSA-N | |
1-(5-bromo-2-hydroxyphenyl)propan-1-one |
C9H9BrO2 | |
MFCD00218577 | |
736409 | |
CCC(=O)C1=CC(Br)=CC=C1O |
Specifications
17764-93-1 | |
156°C | |
MFCD00218577 | |
NDXLYRKTHTZXRC-UHFFFAOYSA-N | |
1-(5-bromo-2-hydroxyphenyl)propan-1-one | |
736409 | |
≥98.0% (GC,T) | |
5′-Bromo-2′-hydroxypropiophenone |
77°C | |
C9H9BrO2 | |
1 g | |
CCC(=O)C1=CC(Br)=CC=C1O | |
229.07 | |
229.07 | |
Crystalline Powder |
Safety and Handling
TSCA : No