missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside, Thermo Scientific Chemicals
Substrate for beta-galactosidase
Supplier: Thermo Scientific Chemicals J60151MD
Description
Substrate for beta-galactosidase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside | |
White | |
MFCD00063780 | |
x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside | |
OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O | |
408.63 | |
CHEBI:75506 | |
Powder |
107021-38-5 | |
C14H15BrClNO6 | |
250 mg | |
OPIFSICVWOWJMJ-UHFFFAOYNA-N | |
2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
10173103 | |
408.63 |
Chemical Identifiers
107021-38-5 | |
408.63 | |
OPIFSICVWOWJMJ-UHFFFAOYNA-N | |
10173103 | |
OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O |
C14H15BrClNO6 | |
MFCD00063780 | |
x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside | |
CHEBI:75506 |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only