missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside, Thermo Scientific Chemicals
Substrate for beta-galactosidase
$442.68 - $839.01
Chemical Identifiers
CAS | 107021-38-5 |
---|---|
Molecular Formula | C14H15BrClNO6 |
Molecular Weight (g/mol) | 408.63 |
MDL Number | MFCD00063780 |
InChI Key | OPIFSICVWOWJMJ-UHFFFAOYNA-N |
Synonym | x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside |
PubChem CID | 10173103 |
ChEBI | CHEBI:75506 |
SMILES | OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ60151MD
|
Thermo Scientific Chemicals
J60151MD |
250 mg |
Each for $442.68
|
|
|||||
AAJ6015103
|
Thermo Scientific Chemicals
J6015103 |
1 g |
Each for $839.01
|
|
|||||
Description
Substrate for beta-galactosidase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107021-38-5 | |
408.63 | |
OPIFSICVWOWJMJ-UHFFFAOYNA-N | |
10173103 | |
OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O |
C14H15BrClNO6 | |
MFCD00063780 | |
x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside | |
CHEBI:75506 |
Specifications
107021-38-5 | |
C14H15BrClNO6 | |
250 mg | |
OPIFSICVWOWJMJ-UHFFFAOYNA-N | |
2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
10173103 | |
408.63 | |
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside |
White | |
MFCD00063780 | |
x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside | |
OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O | |
408.63 | |
CHEBI:75506 | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only