missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215000180
Specifications
5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside | |
7240-90-6 | |
0.994 | |
MFCD00005666 | |
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br | |
408.629 | |
CHEBI:75055 | |
Crystalline Powder |
For HPLC analysis | |
White | |
C14H15BrClNO6 | |
100 mg | |
OPIFSICVWOWJMJ-AEOCFKNESA-N | |
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
65181 | |
408.64 |
Chemical Identifiers
7240-90-6 | |
408.629 | |
OPIFSICVWOWJMJ-AEOCFKNESA-N | |
65181 | |
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
C14H15BrClNO6 | |
MFCD00005666 | |
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
CHEBI:75055 | |
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
Safety and Handling
EINECSNumber : 230-640-8
Recommended Storage : Store at -20°C, desiccate, protect from light .