missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, MP Biomedicals™
$81.01 - $712.38
Chemical Identifiers
CAS | 7240-90-6 |
---|---|
Molecular Formula | C14H15BrClNO6 |
Molecular Weight (g/mol) | 408.629 |
MDL Number | MFCD00005666 |
InChI Key | OPIFSICVWOWJMJ-AEOCFKNESA-N |
Synonym | x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal |
PubChem CID | 65181 |
ChEBI | CHEBI:75055 |
IUPAC Name | (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15000105
|
MP Biomedicals Inc
0215000105 |
5 mg |
Each for $81.01
|
|
|||||
ICN15000150
|
MP Biomedicals Inc
0215000150 |
50 mg |
Each for $174.26
|
|
|||||
ICN15000180
|
MP Biomedicals Inc
0215000180 |
100 mg |
Each for $195.72
|
|
|||||
ICN15000190
|
MP Biomedicals Inc
0215000190 |
500 mg |
Each for $712.38
|
|
|||||
Chemical Identifiers
7240-90-6 | |
408.629 | |
OPIFSICVWOWJMJ-AEOCFKNESA-N | |
65181 | |
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
C14H15BrClNO6 | |
MFCD00005666 | |
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
CHEBI:75055 | |
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
Specifications
7240-90-6 | |
0.994 | |
MFCD00005666 | |
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br | |
408.629 | |
CHEBI:75055 | |
Crystalline Powder |
White | |
C14H15BrClNO6 | |
5 mg | |
OPIFSICVWOWJMJ-AEOCFKNESA-N | |
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
65181 | |
408.64 | |
5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, For HPLC analysis |
Safety and Handling
EINECSNumber : 230-640-8
Recommended Storage : Store at -20°C, desiccate, protect from light .