missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloropentyl acetate, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1111118
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Chloropentyl acetate | |
1.061 | |
97°C (206°F) | |
1.438 | |
50 g | |
5-chloro-n-amyl acetate, 1-acetoxy-5-chloropentane, 5-chloro-1-pentanol acetate, 5-chloro-1-amyl acetate, 5-chloro-1-acetoxypentane, 1-pentanol, 5-chloro-, acetate, 5-chloro-1-pentyl acetate, 1-pentanol, 5-chloro-, 1-acetate, 5-chloroamyl acetate, acetic acid 5-chloro-pentyl ester | |
CC(=O)OCCCCCCl | |
164.629 | |
164.63 |
20395-28-2 | |
96°C to 98°C (11 mmHg) | |
C7H13ClO2 | |
MFCD00013696 | |
1752107 | |
ZCYVIAZIVJNAMO-UHFFFAOYSA-N | |
5-chloropentyl acetate | |
88521 | |
98% |
Chemical Identifiers
20395-28-2 | |
164.629 | |
ZCYVIAZIVJNAMO-UHFFFAOYSA-N | |
88521 | |
CC(=O)OCCCCCCl |
C7H13ClO2 | |
MFCD00013696 | |
5-chloro-n-amyl acetate, 1-acetoxy-5-chloropentane, 5-chloro-1-pentanol acetate, 5-chloro-1-amyl acetate, 5-chloro-1-acetoxypentane, 1-pentanol, 5-chloro-, acetate, 5-chloro-1-pentyl acetate, 1-pentanol, 5-chloro-, 1-acetate, 5-chloroamyl acetate, acetic acid 5-chloro-pentyl ester | |
5-chloropentyl acetate |
Safety and Handling
EINECSNumber : 243-784-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only