missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloropentyl acetate, 90+%, Thermo Scientific Chemicals
$93.51 - $289.75
Chemical Identifiers
CAS | 20395-28-2 |
---|---|
Molecular Formula | C7H13ClO2 |
Molecular Weight (g/mol) | 164.629 |
MDL Number | MFCD00013696 |
InChI Key | ZCYVIAZIVJNAMO-UHFFFAOYSA-N |
Synonym | 5-chloro-n-amyl acetate, 1-acetoxy-5-chloropentane, 5-chloro-1-pentanol acetate, 5-chloro-1-amyl acetate, 5-chloro-1-acetoxypentane, 1-pentanol, 5-chloro-, acetate, 5-chloro-1-pentyl acetate, 1-pentanol, 5-chloro-, 1-acetate, 5-chloroamyl acetate, acetic acid 5-chloro-pentyl ester |
PubChem CID | 88521 |
IUPAC Name | 5-chloropentyl acetate |
SMILES | CC(=O)OCCCCCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1111109
|
Thermo Scientific Chemicals
A1111109 |
10 g |
Each for $93.51
|
|
|||||
AAA1111118
|
Thermo Scientific Chemicals
A1111118 |
50 g |
Each for $289.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20395-28-2 | |
164.629 | |
ZCYVIAZIVJNAMO-UHFFFAOYSA-N | |
88521 | |
CC(=O)OCCCCCCl |
C7H13ClO2 | |
MFCD00013696 | |
5-chloro-n-amyl acetate, 1-acetoxy-5-chloropentane, 5-chloro-1-pentanol acetate, 5-chloro-1-amyl acetate, 5-chloro-1-acetoxypentane, 1-pentanol, 5-chloro-, acetate, 5-chloro-1-pentyl acetate, 1-pentanol, 5-chloro-, 1-acetate, 5-chloroamyl acetate, acetic acid 5-chloro-pentyl ester | |
5-chloropentyl acetate |
Specifications
20395-28-2 | |
96°C to 98°C (11 mmHg) | |
C7H13ClO2 | |
MFCD00013696 | |
1752107 | |
ZCYVIAZIVJNAMO-UHFFFAOYSA-N | |
5-chloropentyl acetate | |
88521 | |
98% |
1.061 | |
97°C (206°F) | |
1.438 | |
10 g | |
5-chloro-n-amyl acetate, 1-acetoxy-5-chloropentane, 5-chloro-1-pentanol acetate, 5-chloro-1-amyl acetate, 5-chloro-1-acetoxypentane, 1-pentanol, 5-chloro-, acetate, 5-chloro-1-pentyl acetate, 1-pentanol, 5-chloro-, 1-acetate, 5-chloroamyl acetate, acetic acid 5-chloro-pentyl ester | |
CC(=O)OCCCCCCl | |
164.629 | |
164.63 | |
5-Chloropentyl acetate |
Safety and Handling
EINECSNumber : 243-784-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only