missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluoro-2-methylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1375003

 View more versions of this product

Catalog No. AAL1375003


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Fluoro-2-methylbenzoyl chloride
1.27
C8H6ClFO
MFCD00236681
UN3265
Moisture sensitive
HBAQRSDZSVESSF-UHFFFAOYSA-N
5-fluoro-2-methylbenzoyl chloride
2737383
98%
21900-39-0
108°C (24 mmHg)
1.532
1 g
2574820
5-fluoro-2-methylbenzoylchloride, 5-fluoro-2-methyl-benzoyl chloride, benzoyl chloride,5-fluoro-2-methyl, acmc-1ciaq, 2-methyl-5-fluorobenzoyl chloride, 5-fluoro-2-methyl benzoylchloride, 5-fluoro-2-methyl benzoyl chloride, benzoyl chloride, 5-fluoro-2-methyl, 5-fluoro-2-methylbenzoyl chloride
CC1=CC=C(F)C=C1C(Cl)=O
172.58
172.59

Chemical Identifiers

21900-39-0
172.58
HBAQRSDZSVESSF-UHFFFAOYSA-N
2737383
CC1=CC=C(F)C=C1C(Cl)=O
C8H6ClFO
MFCD00236681
5-fluoro-2-methylbenzoylchloride, 5-fluoro-2-methyl-benzoyl chloride, benzoyl chloride,5-fluoro-2-methyl, acmc-1ciaq, 2-methyl-5-fluorobenzoyl chloride, 5-fluoro-2-methyl benzoylchloride, 5-fluoro-2-methyl benzoyl chloride, benzoyl chloride, 5-fluoro-2-methyl, 5-fluoro-2-methylbenzoyl chloride
5-fluoro-2-methylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only