Learn More
5-Fluoro-2-methylbenzoyl chloride, 98%, Thermo Scientific Chemicals
$93.75 - $333.74
Chemical Identifiers
CAS | 21900-39-0 |
---|---|
Molecular Formula | C8H6ClFO |
Molecular Weight (g/mol) | 172.58 |
MDL Number | MFCD00236681 |
InChI Key | HBAQRSDZSVESSF-UHFFFAOYSA-N |
Synonym | 5-fluoro-2-methylbenzoylchloride, 5-fluoro-2-methyl-benzoyl chloride, benzoyl chloride,5-fluoro-2-methyl, acmc-1ciaq, 2-methyl-5-fluorobenzoyl chloride, 5-fluoro-2-methyl benzoylchloride, 5-fluoro-2-methyl benzoyl chloride, benzoyl chloride, 5-fluoro-2-methyl, 5-fluoro-2-methylbenzoyl chloride |
PubChem CID | 2737383 |
IUPAC Name | 5-fluoro-2-methylbenzoyl chloride |
SMILES | CC1=CC=C(F)C=C1C(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1375003
|
Thermo Scientific Chemicals
L1375003 |
1 g |
Each for $93.75
|
|
|||||
AAL1375006
|
Thermo Scientific Chemicals
L1375006 |
5 g |
Each for $333.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21900-39-0 | |
172.58 | |
HBAQRSDZSVESSF-UHFFFAOYSA-N | |
2737383 | |
CC1=CC=C(F)C=C1C(Cl)=O |
C8H6ClFO | |
MFCD00236681 | |
5-fluoro-2-methylbenzoylchloride, 5-fluoro-2-methyl-benzoyl chloride, benzoyl chloride,5-fluoro-2-methyl, acmc-1ciaq, 2-methyl-5-fluorobenzoyl chloride, 5-fluoro-2-methyl benzoylchloride, 5-fluoro-2-methyl benzoyl chloride, benzoyl chloride, 5-fluoro-2-methyl, 5-fluoro-2-methylbenzoyl chloride | |
5-fluoro-2-methylbenzoyl chloride |
Specifications
21900-39-0 | |
108°C (24 mmHg) | |
1.532 | |
1 g | |
2574820 | |
5-fluoro-2-methylbenzoylchloride, 5-fluoro-2-methyl-benzoyl chloride, benzoyl chloride,5-fluoro-2-methyl, acmc-1ciaq, 2-methyl-5-fluorobenzoyl chloride, 5-fluoro-2-methyl benzoylchloride, 5-fluoro-2-methyl benzoyl chloride, benzoyl chloride, 5-fluoro-2-methyl, 5-fluoro-2-methylbenzoyl chloride | |
CC1=CC=C(F)C=C1C(Cl)=O | |
172.58 | |
172.59 | |
5-Fluoro-2-methylbenzoyl chloride |
1.27 | |
C8H6ClFO | |
MFCD00236681 | |
UN3265 | |
Moisture sensitive | |
HBAQRSDZSVESSF-UHFFFAOYSA-N | |
5-fluoro-2-methylbenzoyl chloride | |
2737383 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only