missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Formyl-4-methylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2594706

 View more versions of this product

Catalog No. AAH2594706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Formyl-4-methylthiophene-2-boronic acid
118°C to 120°C
MFCD03093890
Air Sensitive
QEIXJCOHLUKRPO-UHFFFAOYSA-N
(5-formyl-4-methylthiophen-2-yl)boronic acid
46738927
97%
352530-25-7
C6H7BO3S
5 g
5-formyl-4-methylthiophen-2-yl boronic acid, 5-borono-3-methylthiophene-2-carboxaldehyde, 5-formyl-4-methylthiophene-2-boronic acid, zlchem 1324, acmc-209iey, 5-borono-3-methylthiophene-2-carbaldehyde, b-5-formyl-4-methyl-2-thienyl boronic acid, boronicacid,b-5-formyl-4-methyl-2-thienyl
CC1=C(SC(=C1)B(O)O)C=O
169.99
169.95

Chemical Identifiers

352530-25-7
169.99
QEIXJCOHLUKRPO-UHFFFAOYSA-N
46738927
CC1=C(SC(=C1)B(O)O)C=O
C6H7BO3S
MFCD03093890
5-formyl-4-methylthiophen-2-yl boronic acid, 5-borono-3-methylthiophene-2-carboxaldehyde, 5-formyl-4-methylthiophene-2-boronic acid, zlchem 1324, acmc-209iey, 5-borono-3-methylthiophene-2-carbaldehyde, b-5-formyl-4-methyl-2-thienyl boronic acid, boronicacid,b-5-formyl-4-methyl-2-thienyl
(5-formyl-4-methylthiophen-2-yl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only