missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Formyl-4-methylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

$172.03 - $500.37

Chemical Identifiers

CAS 352530-25-7
Molecular Formula C6H7BO3S
Molecular Weight (g/mol) 169.99
MDL Number MFCD03093890
InChI Key QEIXJCOHLUKRPO-UHFFFAOYSA-N
Synonym 5-formyl-4-methylthiophen-2-yl boronic acid, 5-borono-3-methylthiophene-2-carboxaldehyde, 5-formyl-4-methylthiophene-2-boronic acid, zlchem 1324, acmc-209iey, 5-borono-3-methylthiophene-2-carbaldehyde, b-5-formyl-4-methyl-2-thienyl boronic acid, boronicacid,b-5-formyl-4-methyl-2-thienyl
PubChem CID 46738927
IUPAC Name (5-formyl-4-methylthiophen-2-yl)boronic acid
SMILES CC1=C(SC(=C1)B(O)O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2594703
View Documents
Thermo Scientific Chemicals
H2594703
1 g
Each for $172.03
 
AAH2594706
View Documents
Thermo Scientific Chemicals
H2594706
5 g
Each for $500.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

352530-25-7
169.99
QEIXJCOHLUKRPO-UHFFFAOYSA-N
46738927
CC1=C(SC(=C1)B(O)O)C=O
C6H7BO3S
MFCD03093890
5-formyl-4-methylthiophen-2-yl boronic acid, 5-borono-3-methylthiophene-2-carboxaldehyde, 5-formyl-4-methylthiophene-2-boronic acid, zlchem 1324, acmc-209iey, 5-borono-3-methylthiophene-2-carbaldehyde, b-5-formyl-4-methyl-2-thienyl boronic acid, boronicacid,b-5-formyl-4-methyl-2-thienyl
(5-formyl-4-methylthiophen-2-yl)boronic acid

Specifications

352530-25-7
C6H7BO3S
1 g
5-formyl-4-methylthiophen-2-yl boronic acid, 5-borono-3-methylthiophene-2-carboxaldehyde, 5-formyl-4-methylthiophene-2-boronic acid, zlchem 1324, acmc-209iey, 5-borono-3-methylthiophene-2-carbaldehyde, b-5-formyl-4-methyl-2-thienyl boronic acid, boronicacid,b-5-formyl-4-methyl-2-thienyl
CC1=C(SC(=C1)B(O)O)C=O
169.99
169.95
5-Formyl-4-methylthiophene-2-boronic acid
118°C to 120°C
MFCD03093890
Air Sensitive
QEIXJCOHLUKRPO-UHFFFAOYSA-N
(5-formyl-4-methylthiophen-2-yl)boronic acid
46738927
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only